Anti-ABCD1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-542
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ABCD1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P33897
- Antigen Synonyms:ABC42|Abcd1|ABCD1_HUMAN|Adrenoleukodystrophy protein|ALD|Aldgh|ALDP|AMN|ATP binding cassette|sub family D (ALD)|member 1|ATP-binding cassette sub-family D member 1|OTTHUMP00000025960|OTTMUSP00000019283|RGD1562128|RP23 373N8.2|X linked adrenoleukodystrophy (ALD) gene homolog
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP-binding cassette sub-family D member 1 protein (632-745AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ABCD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human