Anti-Ly6a Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: Ly6a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77849-602
- 77849-603
- Antigen Symbol:
- Ly6a
- Ly6a
- Antigen Synonyms:
- Ly-6A.2/Ly-6E.|Ly-6A.2/Ly-6E.1|LY6A_MOUSE|Ly6al|Lymphocyte antigen 6A-2/6E-1|Lymphocyte antigen Ly 6A.2/Ly 6E.1|SCA-1|Stem cell antigen 1|T cell activating protein|T-cell-activating protein|TAP
- Ly-6A.2/Ly-6E.|Ly-6A.2/Ly-6E.1|LY6A_MOUSE|Ly6al|Lymphocyte antigen 6A-2/6E-1|Lymphocyte antigen Ly 6A.2/Ly 6E.1|SCA-1|Stem cell antigen 1|T cell activating protein|T-cell-activating protein|TAP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05533
- P05533
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Mouse Ly6a
- Synthetic peptide of Mouse Ly6a
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together