Anti-MED16 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-102
- 77847-101
- Antigen Symbol:
- MED16
- MED16
- Antigen Synonyms:
- DRIP 92|DRIP92|MED 16|MED16|MED16_HUMAN|Mediator complex subunit 16|Mediator of RNA polymerase II transcription subunit 16|THRAP 5|Thyroid hormone receptor associated protein 5|Thyroid hormone receptor associated protein 95 kD subunit|Thyroid hormone receptor associated protein complex 95 kDa component|Thyroid hormone receptor-associated protein 5|Thyroid hormone receptor-associated protein complex 95 kDa component|Trap 95|Trap95|Vitamin D3 receptor interacting protein complex component|Vitamin D3 receptor-interacting protein complex 92 kDa component
- DRIP 92|DRIP92|MED 16|MED16|MED16_HUMAN|Mediator complex subunit 16|Mediator of RNA polymerase II transcription subunit 16|THRAP 5|Thyroid hormone receptor associated protein 5|Thyroid hormone receptor associated protein 95 kD subunit|Thyroid hormone receptor associated protein complex 95 kDa component|Thyroid hormone receptor-associated protein 5|Thyroid hormone receptor-associated protein complex 95 kDa component|Trap 95|Trap95|Vitamin D3 receptor interacting protein complex component|Vitamin D3 receptor-interacting protein complex 92 kDa component
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2X0
- Q9Y2X0
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MED16
- Fusion protein of Human MED16
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together