Anti-RASAL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:20 to 1:100
Type:
Antigen: RASAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-231
- 77847-230
- Antigen Symbol:
- RASAL1
- RASAL1
- Antigen Synonyms:
- GAP1 like protein|MRASAL|RAS GTPase activating protein like|RAS protein activator like 1 (GAP1 like)|RAS protein activator like 1|RASAL|rasGAP activating like protein 1|RasGAP-activating-like protein 1|RASGAP1 LIKE|RASL1_HUMAN
- GAP1 like protein|MRASAL|RAS GTPase activating protein like|RAS protein activator like 1 (GAP1 like)|RAS protein activator like 1|RASAL|rasGAP activating like protein 1|RasGAP-activating-like protein 1|RASGAP1 LIKE|RASL1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95294
- O95294
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human RASAL1
- Fusion protein of Human RASAL1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together