Order Entry
United States
ContactUsLinkComponent
Anti-VPS29 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-VPS29 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-VPS29 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77843-764
  • 77843-763
  • Antigen Symbol:
  • VPS29
  • VPS29
  • Antigen Synonyms:
  • DC15|DC7|DKFZp564F0223|FLJ20492|hVPS29|PEP11|PEP11 homolog|Retromer protein|Vacuolar protein sorting 29|Vacuolar protein sorting 29 (yeast homolog)|Vacuolar protein sorting 29 (yeast)|Vacuolar protein sorting 29 homolog (S. cerevisiae)|Vacuolar protein sorting 29 homolog|Vacuolar protein sorting associated protein 29|Vacuolar protein sorting-associated protein 29|Vacuolar sorting protein VPS29/PEP11|Vesicle protein sorting 29|VPS 29|VPS29|VPS29 retromer complex component|VPS29_HUMAN|x 007 protein
  • DC15|DC7|DKFZp564F0223|FLJ20492|hVPS29|PEP11|PEP11 homolog|Retromer protein|Vacuolar protein sorting 29|Vacuolar protein sorting 29 (yeast homolog)|Vacuolar protein sorting 29 (yeast)|Vacuolar protein sorting 29 homolog (S. cerevisiae)|Vacuolar protein sorting 29 homolog|Vacuolar protein sorting associated protein 29|Vacuolar protein sorting-associated protein 29|Vacuolar sorting protein VPS29/PEP11|Vesicle protein sorting 29|VPS 29|VPS29|VPS29 retromer complex component|VPS29_HUMAN|x 007 protein
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UBQ0
  • Q9UBQ0
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Vacuolar protein sorting-associated protein 29 protein (68-145AA)
  • Recombinant Human Vacuolar protein sorting-associated protein 29 protein (68-145AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together