Anti-CRYBA1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-604
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRYBA1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P05813
- Antigen Synonyms:Beta crystallin A3 isoform A1 Delta4 form|Beta crystallin A3|Beta crystallin A3 isoform A1 Delta7 form|Beta crystallin A3 isoform A1 Delta8 form|Beta-crystallin A3|CRBA1_HUMAN|CRYB1|Crystallin beta A1|Crystallin beta A3|Delta8 form|Eye lens structural protein|isoform A1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-crystallin A3 protein (1-215AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CRYBA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human