Anti-TRIM3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: TRIM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77849-307
- 77849-308
- Antigen Symbol:
- TRIM3
- TRIM3
- Antigen Synonyms:
- BERP|Brain expressed ring finger|Brain-expressed RING finger protein|FLJ16135|HAC1|RING finger protein 22|RING finger protein 97|RNF22|RNF97|TRIM3_HUMAN|Tripartite motif containing 3|Tripartite motif protein Tripartite motif-containing protein 3
- BERP|Brain expressed ring finger|Brain-expressed RING finger protein|FLJ16135|HAC1|RING finger protein 22|RING finger protein 97|RNF22|RNF97|TRIM3_HUMAN|Tripartite motif containing 3|Tripartite motif protein Tripartite motif-containing protein 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75382
- O75382
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TRIM3
- Synthetic peptide of Human TRIM3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together