Anti-RAD18 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAD18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-479
- 77843-478
- Antigen Symbol:
- RAD18
- RAD18
- Antigen Synonyms:
- 2810024C04Rik|DNA repair protein rad18|E3 ubiquitin-protein ligase RAD18|EC 6.3.2.-|hHR 18|hHR18|hRAD 18|hRAD18|MGC156682|Postreplication repair E3 ubiquitin-protein ligase RAD18|Postreplication repair protein hRAD18p|Postreplication repair protein RAD18|RAD 18|RAD18 homolog (S. cerevisiae)|RAD18 homolog|RAD18 S. cerevisiae homolog|RAD18 S. cerevisiae homolog of|RAD18 transcript variant|RAD18_HUMAN|Rad18sc|Radiation sensitivity protein 18|RING finger protein 73|RNF 73|RNF73|Structural maintenance of chromosomes protein 6|YCR066W|YCR66W
- 2810024C04Rik|DNA repair protein rad18|E3 ubiquitin-protein ligase RAD18|EC 6.3.2.-|hHR 18|hHR18|hRAD 18|hRAD18|MGC156682|Postreplication repair E3 ubiquitin-protein ligase RAD18|Postreplication repair protein hRAD18p|Postreplication repair protein RAD18|RAD 18|RAD18 homolog (S. cerevisiae)|RAD18 homolog|RAD18 S. cerevisiae homolog|RAD18 S. cerevisiae homolog of|RAD18 transcript variant|RAD18_HUMAN|Rad18sc|Radiation sensitivity protein 18|RING finger protein 73|RNF 73|RNF73|Structural maintenance of chromosomes protein 6|YCR066W|YCR66W
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase RAD18 protein (1-495AA)
- Recombinant Human E3 ubiquitin-protein ligase RAD18 protein (1-495AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together