Anti-RPH3AL Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RPH3AL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-663
- 77844-662
- Antigen Symbol:
- RPH3AL
- RPH3AL
- Antigen Synonyms:
- Gm1753|No C2 domains protein|Noc2|Rab effector Noc2|Rabphilin 3A-like (without C2 domains)|Rabphilin 3A-like|Rabphilin-3A-like protein|RPH3L_HUMAN
- Gm1753|No C2 domains protein|Noc2|Rab effector Noc2|Rabphilin 3A-like (without C2 domains)|Rabphilin 3A-like|Rabphilin-3A-like protein|RPH3L_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNE2
- Q9UNE2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rab effector Noc2 protein (1-315AA)
- Recombinant Human Rab effector Noc2 protein (1-315AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together