Anti-B3GNT5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-753
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:B3GNT5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BYG0
- Antigen Synonyms:B3GNT5Lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase|EC 2.4.1.206|Lactotriaosylceramide synthase|Lc(3)Cer synthase|Lc3 synthase|UDP-GlcNAc:beta-Gal beta-1,3-N-acetylglucosaminyltransferase 5|BGnT-5|Beta-1,3-Gn-T5|Beta-1,3-N-acetylglucosaminyltransferase 5|Beta3Gn-T5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase protein (65-344AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human