Anti-PORCN Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: PORCN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-091
- 77843-092
- Antigen Symbol:
- PORCN
- PORCN
- Antigen Synonyms:
- DHOF|FODH|MG61|MGC29687|por|PORC|PORCN|PORCN_HUMAN|PPN|Probable protein-cysteine N-palmitoyltransferase porcupine|Protein MG61
- DHOF|FODH|MG61|MGC29687|por|PORC|PORCN|PORCN_HUMAN|PPN|Probable protein-cysteine N-palmitoyltransferase porcupine|Protein MG61
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H237
- Q9H237
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein-serine O-palmitoleoyltransferase porcupine protein (269-332AA)
- Recombinant Human Protein-serine O-palmitoleoyltransferase porcupine protein (269-332AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together