Anti-CHM Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-075
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHM
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P24386
- Antigen Synonyms:dy| Chm| Choroideraemia protein| Choroideremia| DXS540| FLJ38564| GGTA| HSD 32| MGC102710| Rab escort protein 1| Rab geranylgeranyltransferase component A| Rab proteins geranylgeranyltransferase component A 1| RAE1_HUMAN| REP 1| REP-1| REP1| TCD| TCD protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Rab proteins geranylgeranyltransferase component A 1 protein (1-653AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CHM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human