Anti-GDF5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-894
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GDF5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P43026
- Antigen Synonyms:BMP14|Cartilage derived morphogenetic protein 1|Cartilage-derived morphogenetic protein 1|CDMP-1|CDMP1|GDF-5|GDF5_HUMAN|Growth differentiation factor 5|Growth/differentiation factor 5|LAP4|OS5|Radotermin|SYNS2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Growth/differentiation factor 5 protein (202-501AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:20 to 1:200
Type:
Antigen: GDF5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human