Anti-EPS8L2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: EPS8L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-201
- 77843-200
- Antigen Symbol:
- EPS8L2
- EPS8L2
- Antigen Synonyms:
- Epidermal growth factor receptor kinase substrate 8 like protein 2|Epidermal growth factor receptor kinase substrate 8-like protein 2|Epidermal growth factor receptor pathway substrate 8 like protein 2|Epidermal growth factor receptor pathway substrate 8 related protein 2|Epidermal growth factor receptor pathway substrate 8-related protein 2|EPS8 like 2|EPS8 like protein 2|EPS8 related protein 2|EPS8-like protein 2|EPS8-related protein 2|EPS8L 2|EPS8R 2|EPS8R2|ES8L2_HUMAN|FLJ16738|FLJ21935|FLJ22171|MGC126530|MGC3088
- Epidermal growth factor receptor kinase substrate 8 like protein 2|Epidermal growth factor receptor kinase substrate 8-like protein 2|Epidermal growth factor receptor pathway substrate 8 like protein 2|Epidermal growth factor receptor pathway substrate 8 related protein 2|Epidermal growth factor receptor pathway substrate 8-related protein 2|EPS8 like 2|EPS8 like protein 2|EPS8 related protein 2|EPS8-like protein 2|EPS8-related protein 2|EPS8L 2|EPS8R 2|EPS8R2|ES8L2_HUMAN|FLJ16738|FLJ21935|FLJ22171|MGC126530|MGC3088
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H6S3
- Q9H6S3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Epidermal growth factor receptor kinase substrate 8-like protein 2 protein (201-500AA)
- Recombinant Human Epidermal growth factor receptor kinase substrate 8-like protein 2 protein (201-500AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together