Anti-CELF2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-791
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CELF2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O95319
- Antigen Synonyms:Bruno-like protein 3|BRUNOL3|Cardiac elav type RNA binding protein|CELF-2|CELF2_HUMAN|CUG BP2|CUG triplet repeat RNA-binding protein 2|CUG-BP- and ETR-3-like factor 2|CUG-BP2|CUGBP Elav like family member 2|CUGBP Elav-like family member 2|CUGBP2|dJ323N1.1|elav type RNA binding protein|ELAV-type RNA-binding protein 3|ETR 3|ETR-3|ETR3|HGNC:2550|hNAPOR|NAPOR 2|NAPOR|Neuroblastoma apoptosis-related RNA-binding protein|Neuroplastoma apoptosis related RNA binding protein 3|RNA binding protein BRUNOL3|RNA-binding protein BRUNOL-3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human CUGBP Elav-like family member 2 protein (291-381AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:200
Type:
Antigen: CELF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human