Anti-ATP6V1D Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-427
- 77846-428
- Antigen Symbol:
- ATP6V1D
- ATP6V1D
- Antigen Synonyms:
- ATP6V1D|ATP6M|VATD|V-type proton ATPase subunit D|V-ATPase subunit D|V-ATPase 28 kDa accessory protein|Vacuolar proton pump subunit D
- ATP6V1D|ATP6M|VATD|V-type proton ATPase subunit D|V-ATPase subunit D|V-ATPase 28 kDa accessory protein|Vacuolar proton pump subunit D
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5K8
- Q9Y5K8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human V-type proton ATPase subunit D protein (1-247AA)
- Recombinant Human V-type proton ATPase subunit D protein (1-247AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together