Anti-CAPZA2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77852-363
New Product
- Antigen Symbol:CAPZA2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P47755
- Antigen Synonyms:CAPPA2|Capping protein (actin filament) muscle Z line|alpha 2|CapZ alpha 2|CapZ alpha-2|CAPZ|CAZA2_HUMAN|F actin capping protein alpha 2 subunit|F actin capping protein subunit alpha 2|F-actin-capping protein subunit alpha-2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human F-actin-capping protein subunit alpha-2 protein (2-286AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CAPZA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human