Anti-HIST1H2AB HEK293F cells Recombinant Monoclonal Antibody [clone: 1H12]
77850-244
New Product
- Antigen Symbol:HIST1H2AB
- Clonality:Recombinant Monoclonal
- Clone:1H12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P04908
- Antigen Synonyms:Histone H2A type 1-B/E, Histone H2A.2, Histone H2A/a, Histone H2A/m, HIST1H2AB, H2AFM, AND, HIST1H2AE, H2AFA
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:30 to 1:200
Type:
Antigen: HIST1H2AB
Clonality: Recombinant Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human