Anti-AKR7A2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-987
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AKR7A2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:O43488
- Antigen Synonyms:AFAR|AFAR1|AFB1 aldehyde reductase 1|AFB1 AR1|AFB1-AR 1|AFB1AR1|Aflatoxin aldehyde reductase|Aflatoxin B1 aldehyde reductase member 2|Aflatoxin beta1 aldehyde reductase|Aiar|AKR7|Akr7a2|Aldo keto reductase family 7|Aldo keto reductase family 7 member A2 aflatoxin aldehyde reductase|Aldo keto reductase family 7 member A2|member A2 (aflatoxin aldehyde reductase)|Aldoketoreductase 7|ARK72_HUMAN|SSA reductase|Succinic semialdehyde reductase
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aflatoxin B1 aldehyde reductase member 2 protein (100-359AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: AKR7A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse