Anti-CAST Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77852-424
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CAST
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P20810
- Antigen Synonyms:BS 17|Calpain inhibitor|Calpastatin|Cast|Heart type calpastatin|ICAL_HUMAN|MGC9402|Sperm BS 17 component|Sperm BS-17 component
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Calpastatin protein (1-667AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CAST
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human