Anti-ABT1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-585
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ABT1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9ULW3
- Antigen Synonyms:ABT1| ABT1_HUMAN| Activator of basal transcription 1| Basal transcriptional activator 1| Basal transcriptional activator| hABT1| HGNC:17369| OTTHUMP00000017935| TATA-binding protein-binding protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Activator of basal transcription 1 protein (197-270AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ABT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse