Anti-ACAA1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-593
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACAA1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P09110
- Antigen Synonyms:3 ketoacyl CoA thiolase peroxisomal|3-ketoacyl-CoA thiolase|ACAA|Acetyl CoA acyltransferase 1|Acetyl Coenzyme A acyltransferase 1|Acetyl-CoA acyltransferase|Acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3 oxoacyl Coenzyme A|Beta ketothiolase|Beta-ketothiolase|Peroxisomal 3 oxoacyl CoA thiolase|Peroxisomal 3 oxoacyl Coenzyme A thiolase|Peroxisomal 3-oxoacyl-CoA thiolase|peroxisomal|PTHIO|THIK_HUMAN|THIO
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human 3-ketoacyl-CoA thiolase, peroxisomal protein (27-331AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ACAA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human