Anti-SULT1A3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-601
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SULT1A3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:P0DMM9
- Antigen Synonyms:STM|Sulfotransferase 1A3|ST1A3|EC 2.8.2.1|Aryl sulfotransferase 1A3/1A4|Catecholamine-sulfating phenol sulfotransferase|HAST3|M-PST|Monoamine-sulfating phenol sulfotransferase|Placental estrogen sulfotransferase|Sulfotransferase 1A3/1A4|Sulfotransferase|monoamine-preferring|Thermolabile phenol sulfotransferase|TL-PST
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Sulfotransferase 1A3 protein (1-295AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:200
Type:
Antigen: SULT1A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse