Anti-FKBPL Rabbit Polyclonal Antibody
Catalog # 77846-909
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FKBPL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UIM3
- Antigen Synonyms:DIR1|FK506 binding protein like|FK506-binding protein-like|FKBPL_HUMAN|NG7|OTTHUMP00000029170|WAF 1/CIP1 stabilizing protein 39|WAF-1/CIP1 stabilizing protein 39|WISP39
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human FKBPL.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: FKBPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat