Anti-SF1 (phospho-Ser82) Rabbit Polyclonal Antibody
Catalog # 77848-465
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q15637
- Antigen Synonyms:ZFM1|ZNF162|Splicing factor 1|Mammalian branch point-binding protein|BBP|mBBP|Transcription factor ZFM1|Zinc finger gene in MEN1 locus|Zinc finger protein 162
- Modification:phospho-Ser82
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of Serine 82(S-P-S(p)-P-E) derived from Human SF1.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100
Type:
Antigen: SF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse