Anti-ARL4D Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-393
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ARL4D
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P49703
- Antigen Synonyms:ADP ribosylation factor 4 like|ADP ribosylation factor like 4D|ADP ribosylation factor like 6|ADP ribosylation factor like protein 4D|ADP ribosylation factor like protein 4L|ADP-ribosylation factor-like protein 4D|ADP-ribosylation factor-like protein 4L|AR L6|ARF 4L|ARF4L|ARL 4D|ARL 6|ARL4D_HUMAN|ARL6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ADP-ribosylation factor-like protein 4D protein (2-201AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human