Anti-PORCN Rabbit Recombinant Polyclonal Antibody
77843-086
New Product
- Antigen Symbol:PORCN
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H237
- Antigen Synonyms:DHOF|FODH|MG61|MGC29687|por|PORC|PORCN_HUMAN|PPN|Probable protein-cysteine N-palmitoyltransferase porcupine|Protein MG61
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Protein-serine O-palmitoleoyltransferase porcupine protein (269-332AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: PORCN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse