Anti-AK1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-953
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AK1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P00568
- Antigen Synonyms:Adenylate kinase 1|Adenylate kinase isoenzyme 1|Adenylate kinase soluble|AK 1|ATP AMP transphosphorylase|ATP-AMP transphosphorylase 1|KAD1|KAD1_HUMAN|Myokinase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenylate kinase isoenzyme 1 protein (1-194AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human