Anti-HIF3A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HIF3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-583
- 77845-584
- Antigen Symbol:
- HIF3A
- HIF3A
- Antigen Synonyms:
- Basic-helix-loop-helix-PAS protein MOP7|bHLHe17|Class E basic helix-loop-helix protein 17|HIF 3A|HIF 3A4|HIF-3-alpha|HIF3 alpha|HIF3-alpha|HIF3-alpha-1|HIF3A_HUMAN|Hypoxia Inducible Factor 3 alpha|Hypoxia inducible factor 3 alpha subunit|Hypoxia inducible factor three alpha|Hypoxia-inducible factor 3-alpha|Inhibitory PAS domain protein|IPAS|Member of PAS protein 7|MOP7|PAS domain-containing protein 7|PASD7
- Basic-helix-loop-helix-PAS protein MOP7|bHLHe17|Class E basic helix-loop-helix protein 17|HIF 3A|HIF 3A4|HIF-3-alpha|HIF3 alpha|HIF3-alpha|HIF3-alpha-1|HIF3A_HUMAN|Hypoxia Inducible Factor 3 alpha|Hypoxia inducible factor 3 alpha subunit|Hypoxia inducible factor three alpha|Hypoxia-inducible factor 3-alpha|Inhibitory PAS domain protein|IPAS|Member of PAS protein 7|MOP7|PAS domain-containing protein 7|PASD7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2N7
- Q9Y2N7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hypoxia-inducible factor 3-alpha protein (516-669AA)
- Recombinant Human Hypoxia-inducible factor 3-alpha protein (516-669AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together