Anti-ARIH2 Rabbit Recombinant Polyclonal Antibody
77851-376
New Product
- Antigen Symbol:ARIH2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O95376
- Antigen Synonyms:HT005|all trans retinoic acid inducible RING finger|Ari 2|Ari-2|ARI2|ARI2_HUMAN|Ariadne 2|Ariadne 2 protein homolog|ariadne homolog 2 (Drosophila)|ariadne homolog 2|Ariadne RBR E3 ubiquitin protein ligase 2|ARIH 2|E3 ubiquitin protein ligase E3 ubiquitin-protein ligase protein ariadne 2 homolog|Protein ariadne-2 homolog|Triad 1|TRIAD1|Triad1 protein|UIP48
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human E3 ubiquitin-protein ligase ARIH2 protein (194-493AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ARIH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human