Anti-ATP6V1D Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATP6V1D
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-430
- 77846-429
- Antigen Symbol:
- ATP6V1D
- ATP6V1D
- Antigen Synonyms:
- ATP6M|VATD|V-type proton ATPase subunit D|V-ATPase subunit D|V-ATPase 28 kDa accessory protein|Vacuolar proton pump subunit D
- ATP6M|VATD|V-type proton ATPase subunit D|V-ATPase subunit D|V-ATPase 28 kDa accessory protein|Vacuolar proton pump subunit D
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5K8
- Q9Y5K8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human V-type proton ATPase subunit D protein (1-247AA)
- Recombinant Human V-type proton ATPase subunit D protein (1-247AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together