Anti-CCL16 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:100 to 1:300
Type:
Antigen: CCL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-184
- 77848-183
- Antigen Symbol:
- CCL16
- CCL16
- Antigen Synonyms:
- C-C motif chemokine 16|CCL16|CCL16_HUMAN|Chemokine (C-C motif) ligand 16|Chemokine CC-4|Chemokine LEC|CKb12|HCC-4|HCC4|IL-10-inducible chemokine|IL10 inducible chemokine|ILINCK|LCC-1|LCC1|Liver CC chemokine 1 precursor|Liver expressed chemokine|Liver-expressed chemokine|LMC|Lymphocyte and monocyte chemoattractant|Monotactin 1|Monotactin-1|MTN-1|Mtn1|NCC-4|NCC4|New CC chemokine 4|SCYA16|SCYL4|Small inducible cytokine A16 precursor|Small inducible cytokine subfamily A (Cys Cys) member 16|Small-inducible cytokine A16
- C-C motif chemokine 16|CCL16|CCL16_HUMAN|Chemokine (C-C motif) ligand 16|Chemokine CC-4|Chemokine LEC|CKb12|HCC-4|HCC4|IL-10-inducible chemokine|IL10 inducible chemokine|ILINCK|LCC-1|LCC1|Liver CC chemokine 1 precursor|Liver expressed chemokine|Liver-expressed chemokine|LMC|Lymphocyte and monocyte chemoattractant|Monotactin 1|Monotactin-1|MTN-1|Mtn1|NCC-4|NCC4|New CC chemokine 4|SCYA16|SCYL4|Small inducible cytokine A16 precursor|Small inducible cytokine subfamily A (Cys Cys) member 16|Small-inducible cytokine A16
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15467
- O15467
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CCL16
- Synthetic peptide of Human CCL16
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together