Anti-CSE1L Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-640
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CSE1L
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P55060
- Antigen Synonyms:CAS|Cellular apoptosis susceptibility protein|Chromosome segregation 1 (yeast homolog) like|Chromosome segregation 1 Like|Chromosome segregation 1 like protein|Chromosome segregation 1-like protein|Chromosome segregation gene CSE1|CSE 1|CSE 1 chromosome segregation 1 like|CSE 1 chromosome segregation 1 like protein|CSE 1L|CSE1|CSE1 chromosome segregation 1 like (yeast)|CSE1 chromosome segregation 1 like|CSE1 chromosome segregation 1 like protein|CSE1L|Exp 2|Exp2|Exportin 2|Exportin-2|Exportin2|Importin alpha re exporter|Importin-alpha re-exporter|MGC117283|MGC130036|MGC130037|XPO 2|XPO2|XPO2_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Exportin-2 protein (713-971AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CSE1L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human