Anti-MED23 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MED23
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-287
- 77844-286
- Antigen Symbol:
- MED23
- MED23
- Antigen Synonyms:
- 130 kDa transcriptional co activator|133 kDa transcriptional co activator|Activator recruited cofactor 130 kDa component|Activator-recruited cofactor 130 kDa component|ARC 130|ARC130|Cofactor required for Sp1 transcriptional activation subunit 3 130kDa|Cofactor required for Sp1 transcriptional activation subunit 3 130kDa mediator|Cofactor required for Sp1 transcriptional activation subunit 3|CRSP 130|CRSP 130 kD subunit|CRSP 133|CRSP 3|CRSP complex subunit 3|CRSP130|CRSP133|CRSP3|CRSP3 protein|DKFZp434H0117|DRIP 130|DRIP130|hSur-2|KIAA1216|MED 23|MED23_HUMAN|Mediator complex subunit 23|Mediator of RNA polymerase II transcription subunit 23|MRT18|Protein sur-2 homolog|SUR 2|SUR2|Transcriptional co activator CRSP 130|Transcriptional co activator CRSP130|Transcriptional coactivator CRSP130|Vitamin D3 receptor interacting protein|Vitamin D3 receptor interacting protein complex 130 kDa component|Vitamin D3 receptor-interacting protein complex 130 kDa component
- 130 kDa transcriptional co activator|133 kDa transcriptional co activator|Activator recruited cofactor 130 kDa component|Activator-recruited cofactor 130 kDa component|ARC 130|ARC130|Cofactor required for Sp1 transcriptional activation subunit 3 130kDa|Cofactor required for Sp1 transcriptional activation subunit 3 130kDa mediator|Cofactor required for Sp1 transcriptional activation subunit 3|CRSP 130|CRSP 130 kD subunit|CRSP 133|CRSP 3|CRSP complex subunit 3|CRSP130|CRSP133|CRSP3|CRSP3 protein|DKFZp434H0117|DRIP 130|DRIP130|hSur-2|KIAA1216|MED 23|MED23_HUMAN|Mediator complex subunit 23|Mediator of RNA polymerase II transcription subunit 23|MRT18|Protein sur-2 homolog|SUR 2|SUR2|Transcriptional co activator CRSP 130|Transcriptional co activator CRSP130|Transcriptional coactivator CRSP130|Vitamin D3 receptor interacting protein|Vitamin D3 receptor interacting protein complex 130 kDa component|Vitamin D3 receptor-interacting protein complex 130 kDa component
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULK4
- Q9ULK4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mediator of RNA polymerase II transcription subunit 23 protein (1014-1368AA)
- Recombinant Human Mediator of RNA polymerase II transcription subunit 23 protein (1014-1368AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together