Anti-PDLIM1 Rabbit Polyclonal Antibody
Catalog # 77847-690
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PDLIM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O00151
- Antigen Synonyms:C terminal LIM domain protein 1|C-terminal LIM domain protein 1|Carboxy terminal LIM domain protein 1|CLIM 1|CLIM1|CLP 36|CLP36|Elfin|Enigma|Enigma homolog|hCLIM 1|hCLIM1|LIM domain protein CLP 36|LIM domain protein CLP-36|LIM domain protein CLP36|PDLI1_HUMAN|PDLIM 1|PDZ and LIM domain 1|PDZ and LIM domain protein 1
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human PDLIM1.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500
Type:
Antigen: PDLIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat