Anti-DDX3X Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77854-079
New Product
- Antigen Symbol:DDX3X
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O00571
- Antigen Synonyms:ATP dependent RNA helicase ATP-dependent RNA helicase CAP Rf|DBX|DDX14|DDX3X_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 3 X linked|DEAD (Asp-Glu-Ala-Asp) box helicase 3|X-linked|DEAD box|DEAD box protein 3|DEAD box protein 3 X-chromosomal|DEAD box X isoform|X isoform|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 3|DEAD/H box 3|Helicase like protein 2|Helicase-like protein 2|HLP2|X-chromosomal
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP-dependent RNA helicase DDX3X protein (2-662AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: DDX3X
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human