Anti-ZNF695 Rabbit Polyclonal Antibody
Catalog # 77849-609
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZNF695
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IW36
- Antigen Synonyms:MGC61907|RP11-551G24.1|SBZF3|Zinc finger protein 695|Zinc finger protein SBZF3|ZN695_HUMAN|ZNF695
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from C-terminal of Human ZNF695.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ZNF695
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human