Anti-CTSL Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-779
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CTSL
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Chlorocebus aethiops
- Size:50 µg
- Form:Liquid
- Gene ID:Q9GKL8
- Antigen Synonyms:CTSL1Cathepsin L1|EC 3.4.22.15|Cathepsin L|Major excreted protein|MEP) [Cleaved into: Cathepsin L1 heavy chain|Cathepsin L1 light chain]
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Chlorocebus aethiops Cathepsin L1 protein (114-333AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CTSL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chlorocebus aethiops