Anti-C1QTNF3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77852-062
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:C1QTNF3
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BXJ4
- Antigen Synonyms:C1ATNF3|C1q and tumor necrosis factor related protein 3|C1QT3_HUMAN|Cartonectin|Collagenous repeat containing sequence of 26 kDa|collagenous repeat-containing sequence 26 kDa protein|Complement C1q tumor necrosis factor related protein 3|Complement C1q tumor necrosis factor-related protein 3|Corcs|CORS 26|Cors|CORS26|CTRP3|FLJ37576|OTTHUMP00000115918|OTTHUMP00000219981|PRO1484|Secretory protein CORS26|UNQ753
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Complement C1q tumor necrosis factor-related protein 3 protein (23-246AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: C1QTNF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human