Anti-CRABP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-520
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRABP2
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P29373
- Antigen Synonyms:Cellular retinoic acid binding protein 2|Cellular retinoic acid binding protein II|Cellular retinoic acid-binding protein 2|Cellular retinoic acid-binding protein II|CRABP II|CRABP-II|Crabp2|CRABPII|RABP2_HUMAN|RBP6|Rretinoic acid-binding protein|cellular|type II
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cellular retinoic acid-binding protein 2 protein (1-138AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CRABP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human