Anti-POLE Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-264
- 77845-263
- Antigen Symbol:
- POLE
- POLE
- Antigen Synonyms:
- CRCS12|DKFZp434F222|DNA polymerase epsilon catalytic subunit A|DNA polymerase epsilon catalytic subunit|DNA polymerase II subunit A|DPOE1_HUMAN|EC 2.7.7.7|FILS|FLJ21434|POLE1|Polymerase (DNA directed) epsilon|polymerase (DNA directed)|epsilon|catalytic subunit
- CRCS12|DKFZp434F222|DNA polymerase epsilon catalytic subunit A|DNA polymerase epsilon catalytic subunit|DNA polymerase II subunit A|DPOE1_HUMAN|EC 2.7.7.7|FILS|FLJ21434|POLE1|Polymerase (DNA directed) epsilon|polymerase (DNA directed)|epsilon|catalytic subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q07864
- Q07864
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA polymerase epsilon catalytic subunit A protein (1175-1311AA)
- Recombinant Human DNA polymerase epsilon catalytic subunit A protein (1175-1311AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together