Anti-MPP6 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: MPP6
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-645
- 77843-644
- Antigen Symbol:
- MPP6
- MPP6
- Antigen Synonyms:
- MAGUK p55 subfamily member 6|MAGUK p55 subfamily member 6 VELI associated MAGUK 1|MAGUK protein p55T|Membrane protein palmitoylated 6|MPP6_HUMAN|p55T|Palmitoylated 6 MAGUK p55 subfamily member 6|PALS2|Protein associated with Lin7 2|VAM-1|VELI associated MAGUK 1|Veli-associated MAGUK 1
- MAGUK p55 subfamily member 6|MAGUK p55 subfamily member 6 VELI associated MAGUK 1|MAGUK protein p55T|Membrane protein palmitoylated 6|MPP6_HUMAN|p55T|Palmitoylated 6 MAGUK p55 subfamily member 6|PALS2|Protein associated with Lin7 2|VAM-1|VELI associated MAGUK 1|Veli-associated MAGUK 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZW5
- Q9NZW5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human MAGUK p55 subfamily member 6 protein (220-420AA)
- Recombinant Human MAGUK p55 subfamily member 6 protein (220-420AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together