Anti-NDUFAF1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFAF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-135
- 77846-134
- Antigen Symbol:
- NDUFAF1
- NDUFAF1
- Antigen Synonyms:
- CGI 65|CGI65|CIA30|CIA30_HUMAN|Complex I intermediate associated protein 30|Complex I intermediate associated protein 30 mitochondrial|Complex I intermediate-associated protein 30|mitochondrial|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 1|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1|NADH:ubiquinone oxidoreductase complex assembly factor 1|NDUFAF 1|Ndufaf1
- CGI 65|CGI65|CIA30|CIA30_HUMAN|Complex I intermediate associated protein 30|Complex I intermediate associated protein 30 mitochondrial|Complex I intermediate-associated protein 30|mitochondrial|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 1|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1|NADH:ubiquinone oxidoreductase complex assembly factor 1|NDUFAF 1|Ndufaf1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y375
- Q9Y375
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Complex I intermediate-associated protein 30, mitochondrial protein (25-327AA)
- Recombinant Human Complex I intermediate-associated protein 30, mitochondrial protein (25-327AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together