Anti-MCM2 Rabbit Recombinant Polyclonal Antibody
77859-582
New Product
- Antigen Symbol:MCM2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:P49736
- Antigen Synonyms:BM28|CCNL 1|CCNL1|CDC like 1|cdc19|CDCL 1|CDCL1|Cell devision cycle like 1|Cyclin like 1|D3S3194|DNA replication licensing factor MCM2|KIAA0030|MCM 2|MCM2|MCM2 minichromosome maintenance deficient 2 mitotin |MCM2 minichromosome maintenance deficient 2 mitotin (S. cerevisiae)|MCM2 minichromosome maintenance deficient 2|mitotin|MCM2_HUMAN|MGC10606|Minichromosome maintenance complex component 2|Minichromosome maintenance deficient 2 (mitotin)|Minichromosome maintenance deficient 2 mitotin |Minichromosome maintenance protein 2|Minichromosome maintenance protein 2 homolog|Mitotin|Nuclear protein BM28|OTTHUMP00000216047|OTTHUMP00000216050
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA replication licensing factor MCM2 protein (2-904AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:5000, IHC:1:500 to 1:1000, IF:1:200 to 1:500
Type:
Antigen: MCM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human