Anti-APOC2 Rabbit Polyclonal Antibody (Biotinylated)
Catalog # 77851-305
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APOC2
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P02655
- Antigen Synonyms:APC 2| APC2| Apo CII| Apo-CII| APOC 2| ApoC II| ApoC-II| APOC2| APOC2 protein| APOC2_HUMAN| ApoCII| Apolipoprotein C II| Apolipoprotein C II precursor| Apolipoprotein C2| ApolipoproteinCII| MGC75082| ProapoC-II| Proapolipoprotein C-II
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence from Human Apolipoprotein C-II protein (20-38AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: APOC2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human