Anti-WNK1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: WNK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-130
- 77843-129
- Antigen Symbol:
- WNK1
- WNK1
- Antigen Synonyms:
- Erythrocyte 65 kDa protein|HSAN2|HSN2|hKDP|KIAA0344|Kinase deficient protein|MGC163339|MGC163341|p65|PPP1R167|PRKProstate derived sterile 20 like kinase|Protein kinase lysine deficient 1|Protein kinase lysine-deficient 1|Protein kinase with no lysine 1|Protein phosphatase 1|regulatory subunit 167|PSK|Serine/threonine protein kinase WNK1 1|Serine/threonine protein kinase WNK1 2|Serine/threonine protein kinase Serine/threonine-protein kinase With no K|WNK lysine deficient protein kinase 1|WNK lysine deficient protein kinase 1 isoform|WNK1_HUMAN
- Erythrocyte 65 kDa protein|HSAN2|HSN2|hKDP|KIAA0344|Kinase deficient protein|MGC163339|MGC163341|p65|PPP1R167|PRKProstate derived sterile 20 like kinase|Protein kinase lysine deficient 1|Protein kinase lysine-deficient 1|Protein kinase with no lysine 1|Protein phosphatase 1|regulatory subunit 167|PSK|Serine/threonine protein kinase WNK1 1|Serine/threonine protein kinase WNK1 2|Serine/threonine protein kinase Serine/threonine-protein kinase With no K|WNK lysine deficient protein kinase 1|WNK lysine deficient protein kinase 1 isoform|WNK1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H4A3
- Q9H4A3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase WNK1 protein (1-207AA)
- Recombinant Human Serine/threonine-protein kinase WNK1 protein (1-207AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together