Anti-FEM1B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FEM1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-412
- 77844-411
- Antigen Symbol:
- FEM1B
- FEM1B
- Antigen Synonyms:
- F1A alpha|F1A-alpha|F1AA|Fem-1-like death receptor-binding protein alpha|Fem-1-like in apoptotic pathway protein alpha|Fem1 homolog b|FEM1 like death receptor binding protein|FEM1-beta|FEM1B_HUMAN|Protein fem-1 homolog B
- F1A alpha|F1A-alpha|F1AA|Fem-1-like death receptor-binding protein alpha|Fem-1-like in apoptotic pathway protein alpha|Fem1 homolog b|FEM1 like death receptor binding protein|FEM1-beta|FEM1B_HUMAN|Protein fem-1 homolog B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UK73
- Q9UK73
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein fem-1 homolog B protein (400-627AA)
- Recombinant Human Protein fem-1 homolog B protein (400-627AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together