Anti-SLC5A9 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: SLC5A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77849-226
- 77849-225
- Antigen Symbol:
- SLC5A9
- SLC5A9
- Antigen Synonyms:
- Solute carrier family 5 member 9|Na(+)/glucose cotransporter 4|Sodium/glucose cotransporter 4|SGLT4|hSGLT4
- Solute carrier family 5 member 9|Na(+)/glucose cotransporter 4|Sodium/glucose cotransporter 4|SGLT4|hSGLT4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q2M3M2
- Q2M3M2
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC5A9
- Synthetic peptide of Human SLC5A9
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together