Anti-HOXA4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-563
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXA4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q00056
- Antigen Synonyms:Dfd like protein|Homeo box A4|Homeobox A4|Homeobox protein Hox-1.4|Homeobox protein Hox-1D|Homeobox protein Hox-A4|Hox 1.4 like protein|hox-1.4|hox-1d|HOX1|HOX1D|HXA4_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Homeobox protein Hox-A4 protein (23-173AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000
Type:
Antigen: HOXA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat