Anti-SLC9A7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: SLC9A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77847-963
- 77847-962
- Antigen Symbol:
- SLC9A7
- SLC9A7
- Antigen Synonyms:
- Na(+)/H(+) exchanger 7|NHE-7|NHE7|Nonselective sodium potassium/proton exchanger|SL9A7_HUMAN|SLC9A6|Sodium/hydrogen exchanger 7|Solute carrier family 9 (sodium/hydrogen exchanger)|member 7|Solute carrier family 9 member 7|Solute carrier family 9|isoform 7
- Na(+)/H(+) exchanger 7|NHE-7|NHE7|Nonselective sodium potassium/proton exchanger|SL9A7_HUMAN|SLC9A6|Sodium/hydrogen exchanger 7|Solute carrier family 9 (sodium/hydrogen exchanger)|member 7|Solute carrier family 9 member 7|Solute carrier family 9|isoform 7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96T83
- Q96T83
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC9A7
- Synthetic peptide of Human SLC9A7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together